Pipette Box, White PVC, Adjustable, TrippNT
Fournisseur: TrippNT
Certificats
A propos de cet article
The inside distance of this storage box is 25" making it perfect for long, large volume, volumetric pipets.
- Stores and separates long pipettes
- Dividers can move to accommodate storage needs
- Choose from 2 sizes
- White PVC construction with optional acrylic lid
- In stock and ready to ship
- American made by TrippNT
- Total replacement Lifetime Guarantee
The hinged lid (which is optional) protects supplies from dust or dirt and extra protection from breakage. You can also change the size of the compartments with the adjustable dividers so it fits exactly the number and size of your items.
Customers Who Bought This Also Bought