Shelf, Rotating Square, PVC, TrippNT
Fournisseur: TrippNT
Certificats
A propos de cet article
These 360 degree rotating shelves have four compartments on each level.
- Divided shelves rotate and sort supplies
- 3 levels
- Heavy-duty, smooth, rotating turntable
- Each level has 4 bins
- Useful in cabinets, refrigerators
- Helpful to share supplies between workstations
- Durable white PVC construction
- American made by TrippNT
- Total replacement Lifetime Guarantee
A safety lip keeps items safely on each 12 x 12 inch square shelf.
Customers Who Bought This Also Bought