Fluff Underpad, DUKAL™ Corporation
Supplier: Dukal
Certificates
About this item
Highly absorbent with a cellulose fill that does not shift during use.
- Soft spunbonded facing and blue polypropylene backing
- Vacuum packed for maximum storage efficiency
- Not made with natural rubber latex
Packaging: Packaged 100 per bag, 3 bags per case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.