VACUETTE® Specimen Collection System, Greiner Bio-One
Safety Holder
Supplier: Greiner Bio-One
Fournisseur #: 450230
Jump to:
Product Details & Documents
The completely assembled VACUETTE® Specimen Collection System is equipped with a rotational protective shield that can be activated with the aid of a solid surface.
- Rotational protective shield prevents needlestick injuries
- Full coverage through rotation provides extra user comfort
- Designed for use with VACUETTE® multiple-sample drawing needles
The protective shield is attached to the holder and, once activated, keeps the needle contained to help prevent needlestick injuries. The rotational shield can rotate 360° to provide user comfort. Holders are designed exclusively for use with VACUETTE® multiple-sample drawing needles or VACUETTE® VISIO PLUS needles. The Specimen Collection System combines a safety tube holder and a VACUETTE® VISIO PLUS needle.
Spécifications
- Désignation:Safety Holder