To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Excitatory amino acid transporter 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:EAAT3
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:10 to 27
- Sequence:DWRRFLRNHWLLLSTVAA
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of mouse EAAT3 (10-27aa DWRRFLRNHWLLLSTVAA), identical to the related rat sequence.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-666
- Supplier No.:PA2164-2
Spécifications
A propos de cet article
Rabbit IgG polyclonal antibody for Excitatory amino acid transporter 3(SLC1A1) detection. Tested with WB in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: EAAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat