To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:FGF1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:aFGF|FGF 1|HBGF-1|FGF1|Acidic fibroblast growth factor|Endothelial cell growth factor alpha|Heparin-binding growth factor 1|Putative heparin-binding growth factor 1|Fibroblast Growth Factor 1|Alpha-endothelial cell growth factor|FGF-1
- Amino acid number:16 to 155
- Storage temperature:–20 °C
- Immunogen:E. coli-derived human FGF1 recombinant protein (Position: F16-D155).
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-326
- Supplier No.:RP1005
Spécifications
A propos de cet article
Rabbit IgG polyclonal antibody for Fibroblast growth factor 1(FGF1) detection. Tested with WB, IHC-P, ELISA, Neu, IP in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: FGF1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human