To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 2
- Antigen symbol:SOX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Antigen synonyms:MCOPS3|ANOP3|SRY-related HMG-box gene 2|transcription factor SOX-2|transcription factor SOX2|sex determining region Y box 2
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human SOX2 (171-183aa YSMMQDQLGYPQH), different from the related mouse and rat sequence by one amino acid.
- Amino acid number:171 to 183
- Purification:purified by immunogen affinity purification
- Sequence:YSMMQDQLGYPQH
- Storage temperature:–20 °C
- Form:Lyophilized
- Cross adsorption:No
- Cat. no.:10207-440
- Supplier No.:PA2284
Specifications
About this item
Rabbit IgG polyclonal antibody for Transcription factor SOX-2(SOX2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat