To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box P3
- Antigen symbol:FOXP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Antigen synonyms:AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human FOXP3 (321-337aa FPEFLHNMDYFKFHNMR), different from the related mouse sequence by two amino acids.
- Amino acid number:321 to 337
- Purification:purified by immunogen affinity purification
- Sequence:FPEFLHNMDYFKFHNMR
- Storage temperature:–20 °C
- Form:Lyophilized
- Cross adsorption:No
- Cat. no.:10206-858
- Supplier No.:PA1577
Specifications
About this item
Rabbit IgG polyclonal antibody for Forkhead box protein P3(FOXP3) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: FOXP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human