J-KEM® Temperature Controller, Model 210, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates
Controller has the ability to display temperature without heating. The Model 210 controls temperature to within ± 0.1 °C of liquids from 1µL to 5L.
Ordering information: Includes 1/8” dia. x 12” long PTFE coated thermocouple, 10 feet coiled extension cord, and a 14/20 PTFE thermocouple adapter.
Customers Who Bought This Also Bought