HyperREZ™ XP Guard Cartridge Holder, Thermo Scientific
# de catalogue 10067-730
Fournisseur: Thermo Scientific
Spécifications
- Description:Guard Cartridge Holder, I.Ø×L: 5.0×5 mm
- Cat. no.:10067-730
- Supplier No.:60002-354
Spécifications
A propos de cet article
Choose the Thermo Scientific HyperREZ XP Guard Cartridge Holder for use with HyperREZ XP 3.0 x 5.0 mm Guard Cartridges.
Customers Who Bought This Also Bought