Uniguard™ Direct-Connection Guard Cartridge Holders, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Eliminate the requirement for extra fittings using Thermo Scientific Uniguard Direct-Connection Guard Cartridge Holders
With PEEK 0.0625" male outlet that fits many columns; 0.0625" female inlet tip can be used with various standard fittings.
They are reusable, stainless steel guard cartridge holders that attach directly to the analytical column inlet.
Customers Who Bought This Also Bought