Lens Cleaning Station, MCR Safety
# de catalogue 80081-970
Fournisseur: MCR Safety
Spécifications
- Description:Lens cleaning station incl. 2 boxes of 300 tissues and one 16-ounce bottle of cleaning spray
- Cat. no.:80081-970
- Supplier No.:LCS1
Spécifications
A propos de cet article
This cleaning station contains 2 boxes of 300 tissues and one 16-ounce bottle of anti-fog, anti-static cleaning spray. It assist in the care and maintenance of your safety eyewear.
- For plastic, PC and glass lenses
- Anti-fog, anti-static formula
- Contains no silicone
Ideal for glasses, goggles, face shields and computer screens.
Customers Who Bought This Also Bought