Rheodyne® Inlet Filters, IDEX Health & Science
# de catalogue 50806-987
Fournisseur: Rheodyne
Spécifications
- Description:0.5 µm replacement filter discs, For 7335
- Cat. no.:50806-987
- Supplier No.:7335-010
Spécifications
A propos de cet article
Inlet filters are used to extract particles from the sample between the injector and the column.
Customers Who Bought This Also Bought