Micro-Splitter Valve, Upchurch Scientific®
# de catalogue 53500-630
Fournisseur: Upchurch Scientific
Spécifications
- Description:Micro-splitter valve, medium pressure, 1/4-28 threads, biocompatible, 2.1 / 4.1 µL (closed / fully open) internal volume
- Cat. no.:53500-630
- Supplier No.:P-450
Spécifications
A propos de cet article
Designed to accurately split and control a low-flow stream of a single incoming supply.
- For interfacing LC-MS systems
- Pressure rated to 800 psi (55 bar)
- Adjustable split stream flow ratesFor interfacing LC-MS systems
- Pressure rated to 800 psi (55 bar)
- Adjustable split stream flow ratesFor interfacing LC-MS systems
- Pressure rated to 800 psi (55 bar)
- Adjustable split stream flow rates
Customers Who Bought This Also Bought