Medium Pressure 4-Port Switching Valves, Upchurch Scientific®, IDEX Health & Science
# de catalogue 53498-576
Fournisseur: Upchurch Scientific
Spécifications
- Description:4-way diagonal flow switching valve, bulkhead version, 1/16" OD tubing, 1.0 mm thru-hole, 12.4 µL swept volume
- Cat. no.:53498-576
- Supplier No.:V-101D
Spécifications
A propos de cet article
This 4 Way PEEK Bulkhead Flow Valve is ideal if your application involves low frequency of use and demands operator control or involves injection of smaller sample volumes.
- Biocompatible and chemically inert
- Pressure rated to 500 psi (34 bar)
Customers Who Bought This Also Bought