Rheodyne® MXP High Pressure Valves, MX Series II™, IDEX Health & Science
Fournisseur: Rheodyne
Certificats
A propos de cet article
Chemically compatible for use with most mobile phase compositions.
- Can withstand up to 6000 psi (414 bar)
- Available with analytical and nano-scale flow paths
- Multiple control options
- Stand alone capability
- Included software
Customers Who Bought This Also Bought