Rheodyne® MXX Low Pressure Valves, MX Series II™, IDEX Health & Science
Fournisseur: Avantor
Certificats
A propos de cet article
Combine MX Series II™ modular valves with an instrument currently in use to support complex fluid switching and sample injection needs.
- Can withstand up to 125 psi (9 bar)
- All models accept 1/16" or 1/8" tubing (proper ferrules required)
- Multiple control options
- Stand alone capability
- Included software
Customers Who Bought This Also Bought