Manual 2-Position Switching Valves, Rheodyne®, IDEX Health & Science
# de catalogue 97058-314
Fournisseur: Rheodyne
Spécifications
- Description:2-position, 6-port switching valve, large bore, 10-32 ports, 1.0 mm stator passage diameter, Stainless steel, For 1/16" OD tubing
- Cat. no.:97058-314
- Supplier No.:7000L
Spécifications
A propos de cet article
Applicable for sample injection, column switching and sample enrichment applications.
Customers Who Bought This Also Bought