Upchurch Scientific® Sample Loops for High Pressure Valves, IDEX Health & Science
# de catalogue 53501-296
Fournisseur: Upchurch Scientific
Spécifications
- Capacity:10000 µl
- Tubing I.D.:2.03 mm
- Tubing O.D.:⅛"
- Description:Sample loop
- Material:Stainless steel
- Cat. no.:53501-296
- For Use With:7125, 7010
- Supplier No.:1876