Digital Electrostatic Field Meter, STATICO
# de catalogue 14223-826
Fournisseur: Statico
Spécifications
- Description:Digital Electrostatic Field Meter
- Cat. no.:14223-826
- Supplier No.:S2001V
Spécifications
A propos de cet article
The S2001 is designed to locate and measure static charge potentials on product, people, equipment and packaging
The instrument uses a non-contacting, chopper stabilized field sensor and a ranging light mechanism to ensure correct measurements of electrostatic fields in all areas- even those using air ionization. The Fieldmeter factory calibrated and can easily be zeroed with the turn of a small knob and does not require re-zeroing between measurements. Powered by a 9 Volt battery, the S2001 features a conductive case and a convenient snap-on ground lead to facilitate grounding and increase accuracy.
Certifications: NIST Certified.
Ordering information: The fieldmeter comes with a 6' Mini Grounding Coil Cord, a 9V Battery, and is housed in a carrying case.
Customers Who Bought This Also Bought