Ferrules, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Thermo Scientific ferrules are available in three different materials and various dimensions to match the instrument and capillary column I.D
Thermo Scientific 100% graphite ferrules are made of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections. These easy-to-use ferrules form a soft grip with the column and provide a stable seal.
The choice of material is dependent upon the use.
Customers Who Bought This Also Bought