Liner Sealing Rings for Thermo Scientific Instruments, Thermo Scientific
Catalog # 10055-060
Supplier: Thermo Scientific
Specifications
- Description:Liner sealing ring
- Cat. no.:10055-060
- For Use With:TRACE GC PTV injector inlet
- Supplier No.:29013417
Specifications
About this item
Liner Sealing Rings made of graphite for Thermo Scientific PTV injectors.
Customers Who Bought This Also Bought