Storage Vials and Closures
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
These storage screw thread vials are manufactured from superior quality 33 expansion borosilicate clear Type 1, Class A or 51A amber Type 1, Class B glass.
- PTFE-lined solid-top storage caps
- Not autoclavable
- PTFE on film/foam packing offers broad chemical resistance
The storage vials range from 2 to 40 ml.
Customers Who Bought This Also Bought