SP Wilmad-LabGlass Ross Miles Foam Test Apparatus, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Designed for testing foaming properties of surface active agents.
- Reference: ASTM D 1173
Foam test apparatus consists of a 200mL foam pipette with a 2mm PTFE straight bore stopcock, and a foam receiver with graduations at 50mL and 250mL above a 6mm PTFE straight bore stopcock.
Customers Who Bought This Also Bought