Three-Layer Static Dissipative Vinyl Mat, STATICO
Supplier: Statico
Certificates
About this item
S1512 Series is a Blue 3-Layer Static Dissipative construction mat that provides a unique work surface for use in anti static workstations to protect ESD (electrostatic discharge) sensitive components and devices against static electricity
The conductive buried fleece helps to increase dimensional stability and lay-flat qualities of the mat. While the top layer provides a tough work surface, the bottom layer adds excellent cushioning properties to the mat. Thickness is 3.18mm (0.125 in.) (nominal).
Certifications: Meets ANSI/ESD S20.20-2014.
Ordering information: Includes one (1) 4.6m (15 ft.) Mat/Wrist Strap Common Point Grounding Cord and two (2) 10mm (3/8 in.) Male Studs installed or un-installed.
Customers Who Bought This Also Bought