DigiTUBE® Digestion Tubes
Fournisseur: PERKINELMER U.S. LLC
Certificats
A propos de cet article
Disposable sample digestion tubes with Class A graduations.
- Easy-to-read graduations
- Allows normalization directly in the tube
- Easy one-handed closure of leak-free screw cap
All-in-one tubes eliminate the need for five separate vessels such as Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.
Customers Who Bought This Also Bought