Stainless Steel Compact Cart System, Micronova
Fournisseur: Micronova
Certificats
A propos de cet article
Compact and highly manoeuvrable stainless steel cart system with smooth, stainless steel buckets and 360° rotation Prevenz Antimicrobial Wheels. Available in single, double and triple bucket configurations.
- Durable bacteria-resistant construction
- Prevenz Antimicrobial Wheels with 360° rotation
- Smooth bucket interiors with no ridges or seams to harbor bacteria
- Compact size allows for easy storage
- Generous width easily accommodates 45.7 cm (18") mops
- Foot activated brake for stability while cleaning
- Bucket includes a stainless steel carrying handle
- Wringers attach directly to rim of bucket
- Optional wringer bar allows W-6 wringer to be positioned above the rinse bucket
- Entire unit is autoclavable
The Micronova Stainless Steel Compact Cart System features a bacteria-resistant construction with antimicrobial wheels, smooth, molded stainless steel buckets with no ridges or seams and a compact, stainless steel frame. Compatible with MegaWringer™ and Slim-T™ series stainless steel mop wringers. The C3A-03 Wringer Bar attachment positions the W-6 Downpress Wringer above the rinse bucket or empty third bucket for simplified wringing.
Ordering information: Buckets are included with carts. All other components are optional and must be ordered separately.
Spécifications
- Catalog No:
- 94000-434
- 68400-672
- 89140-710
- 89423-422
- 89423-424
- 89423-420
- 89423-430
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials, Energy Efficient