SensaTouch™ Gloves, Industry Standard, Powder-Free, MCR Safety
Fournisseur: MCR Safety
Certificats
A propos de cet article
Rubber latex is, by its very nature, a natural substance
- Reversible
- Smooth grip
- Powder free
The natural rubber latex polymer used in the manufacture of natural rubber gloves is a biodegradable material that is inherently antimicrobial and antibacterial. The Sensagaurd latex 5055 features 5 mil thickness, industrial/food service grade. These products contain natural rubber latex which may cause allergic reactions in some people.
Ordering information: These gloves are packaged as 100 per dispenser.
Spécifications
- Catalog No:
- 80081-397
- 80081-395
- 80081-401
- 80081-399
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- FDA regulated:
- +
- +
- +
- +