MedTech Gloves™, Disposable, Medical Grade Gloves, Textured, Powder-Free, NFPA Certified, MCR Safety
Fournisseur: MCR Safety
Certificats
A propos de cet article
Rubber latex is, by its very nature, a natural substance.
- Textured grip
- Powder free
The natural rubber latex polymer used in the manufacture of natural rubber gloves is a biodegradable material that is inherently antimicrobial and antibacterial. The 5049 features 11 mil thickness, is medical grade (Class 1 medical device).
Certifications: NFPA 1999 certified.
Caution: These products contain natural rubber latex which may cause allergic reactions in some people.
Spécifications
- Catalog No:
- 80081-375
- 80081-373
- 80081-377
- 80081-371
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- FDA regulated:
- +
- +
- +
- +