ContactUsLinkComponent
GPI Anatomicals® Canine Knee
GPI Anatomicals® Canine Knee
# de catalogue 470160-276
Fournisseur:  GPI ANATOMICALS
Small Business Enterprise
Model canine knee, size: 2x3-1/2x7IN, Card: 5-1/4x6-1/2IN, Base:6-1/2x5inch
GPI Anatomicals® Canine Knee
# de catalogue 470160-276
Fournisseur:  GPI ANATOMICALS
Small Business Enterprise
Spécifications
  • Description:
    Knee
  • Height:
    18 cm
  • No. of Keyed Items:
    27
  • Cat. no.:
    470160-276
  • Length:
    17 cm
  • Width:
    13 cm
  • Supplier No.:
    9050
  • No. of Parts:
    1
Spécifications
A propos de cet article
These canine joint models are small size illustrations of how these joints function. All relevant bones and tendons are present at each joint.
  • Each model is supplied on a base with a keycard.
Use as part of a comparative anatomy lesson, or in a veterinary situation to explain to a patient how the joint functions.
Customers Who Bought This Also Bought