GPI Anatomicals® Metabolic Syndrome Model
# de catalogue 470160-274
Fournisseur: GPI ANATOMICALS
Small Business Enterprise
Spécifications
- Description:Metabolic Syndrome Model
- Height:29 cm
- No. of Keyed Items:22
- Cat. no.:470160-274
- Length:16 cm
- Width:23 cm
- Supplier No.:4020
- No. of Parts:7
Spécifications
A propos de cet article
This is miniature of brain, heart, kidney, liver, artery, pancreas.
Education card illustrates effects associated with Metabolic Syndrome: stroke, congestive heart failure, coronary artery disease, angina, myocardial infarction, no-alcoholic fatty liver disease, artherosclerosis, renal arteriosclerosis, nephrosclerosis.
Customers Who Bought This Also Bought