GPI Anatomicals® Prostate Disease Model
# de catalogue 470160-256
Fournisseur: GPI ANATOMICALS
Small Business Enterprise
Spécifications
- Description:Prostate Disease Model
- Height:16 cm
- No. of Keyed Items:6
- Cat. no.:470160-256
- Length:23 cm
- Width:16 cm
- Supplier No.:3000
- No. of Parts:1
Spécifications
A propos de cet article
This model is the demonstration of prostate disease model.
- Shows 9 Pathologies
- Keycard Mounted On Base
Complete unit includes six full size prostates. Conditions shown include: normal prostate; normal size prostate with hard nodule below right lobe surface; prostate with enlarged right lobe; enlarged prostate, symmetrical surface, slight median furrow; enlarged prostate, hard nodule below right base surface; enlarged prostate with hard irregular surface and seminal vesicle involvement.
Customers Who Bought This Also Bought