Human TNFSF4/OX40L PicoKine ELISA Kit, Boster
Catalogue # 10207-598
Fournisseur: Boster Biological Technology
Numéro CAS:
Spécifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:TNFSF4/OX40L
- Size:96 well
- Immunogen:Standard: NSO, Q51-L183
- Cat. no.:10207-598
- Supplier No.:EK0857
Spécifications
À propos de cet article
Sandwich ELISA kit of Quantitative Detection for Human TNFSF4/OX40L
Ordering information: Kits include 96-well plate precoated with anti- human TNFSF4 antibody 1; ST0000-10 Lyophilized recombinant human TNFSF4standard 10ng/tubex2; AR1107 Biotinylated anti- human TNFSF4 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.