Rat BMP-2 PicoKine ELISA Kit, Boster
# de catalogue 10205-812
Fournisseur: Boster Biological Technology
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Assay Type:Sandwich
- Reactivity:Rat
- Size:96 well
- Immunogen:Standard: CHO, Q283-R396
- Cat. no.:10205-812
- Supplier No.:EK0312
Spécifications
A propos de cet article
Sandwich ELISA kit of Quantitative Detection for Rat BMP-2
Ordering information: Kits include 96-well plate precoated with anti- rat BMP-2 antibody 1; ST0000-10 Lyophilized recombinant rat BMP-2 standard 10ng/tubex2; AR1107 Biotinylated anti- rat BMP-2 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.
Customers Who Bought This Also Bought