GPI Anatomicals® Osteoarthritis Stages Model
# de catalogue 470160-240
Fournisseur: GPI ANATOMICALS
Small Business Enterprise
Spécifications
- Description:Osteoarthritis Stages Model
- Height:16 cm
- No. of Keyed Items:30
- Cat. no.:470160-240
- Length:27 cm
- Width:16 cm
- Supplier No.:1800
- No. of Parts:4
Spécifications
A propos de cet article
Four smaller shoulder models that depict glenohumeral joint osteoarthritis, a degenerative joint ailment.
- Shows Multiple Effects Of Osteoarthritis
- Removable From Base For Examination
- Keycard Mounted On Base
Set of four shoulder models, reduced size, illustrating degenerative joint disease (osteoarthritis) of glenohumeral joint: erosion to joint articular cartilage, progression of degenerative disease, osteophyte (bone spur) formation at the articular surfaces, and humeral head flattening. Normal stage includes a full scapula. Advanced stage shows acromioclavicular joint osteoarthritis and ligaments.
Customers Who Bought This Also Bought