GPI Anatomicals® Muscled Hip with Sciatic Nerve Model
# de catalogue 470160-230
Fournisseur: GPI ANATOMICALS
Small Business Enterprise
Spécifications
- Description:Muscled Hip with Sciatic Nerve Model
- Height:25 cm
- No. of Keyed Items:26
- Cat. no.:470160-230
- Length:19 cm
- Width:12 cm
- Supplier No.:1311
- No. of Parts:2
Spécifications
A propos de cet article
This model is the demonstration of full size normal right hip with proximal femur and lower lumbar vertebrae.
Full size normal right hip with proximal femur and lower lumbar vertebrae including: sciatic nerve, gluteus medius, gluteus minimus, iliacus, inferior and superior gemellus, obturator internus, piriformis and psoas muscles, articular capsule ligaments, L4-L5 with sacrum.
Customers Who Bought This Also Bought