Ceramic Multibore Tubes, AL23, Round, Thermo Scientific
Fournisseur: Avantor
Certificats
A propos de cet article
Pure Al₂O₃ Aluminium oxide, AL23.
- Corrosion resistant
- Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.