Balance Power Adapters and Wall Plugs
Fournisseur: OHAUS CORP.
Certificats
A propos de cet article
These power cords and wall plugs will power all your Ward's and Ohaus balances.
- Find appropriate power cord/wall plug combo from the table
- Some models require just the power cord
- Some models require a power cord and wall plug
Please use the pictured chart to locate the appropriate combination of power cord and wall plug (if needed) for your specific balance model. Some models require both a power cord and a wall plug, while others only require the power cord. This is indicated in the pictured table.
Customers Who Bought This Also Bought