ContactUsLinkComponent
HSP70 High Sensitivity ELISA Kit, Enzo Life Sciences
HSP70 High Sensitivity ELISA Kit, Enzo Life Sciences
# de catalogue 89141-860
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
HSP70 HIGH SENSITIVITY EIA Kit 1 X 96 Wells
HSP70 High Sensitivity ELISA Kit, Enzo Life Sciences
# de catalogue 89141-860
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sensitivity:
    90 pg/ml (range 0.20-12.5 ng/ml)
  • Target protein:
    HSP70
  • Cat. no.:
    89141-860
  • Supplier No.:
    EKS-715
  • No. of Tests:
    96 Wells
Spécifications
A propos de cet article
Only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples.
For the quantitative determination of HSP70 in serum and plasma of human, mouse, and rat origin. Cited sample(s) include cell lysates and urine.
The HSP70 high sensitivity EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Absorbance is read at 450 nm. This is the only commercially available kit specifically validated and sensitive enough to quantify HSP70 (HSP72) in serum and plasma samples, as referenced by the Cell Stress Society International.
Ordering information: Contains Microtiter plate, Assay buffer 28, Standard, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.
Customers Who Bought This Also Bought