Labels and Tapes "Radioactive Warning", Shamrock
# de catalogue 36436-249
Fournisseur: Shamrock Scientific
Small Business Enterprise
Spécifications
- Description:Warning label tape
- Cat. no.:36436-249
- Legend:Radioactive
- Dimensions:25 mm (1")
- Supplier No.:SRA-2PKG
Spécifications
A propos de cet article
Printed with vivid magenta ink on bright yellow background, labels can be used on any surface where visual warning is required.
Ordering information: Labels are available on rolls of 500 labels. Tape is supplied in 500 rolls.
Customers Who Bought This Also Bought