ContactUsLinkComponent
Cyto-ID® Green Long-Term Cell Tracer Kit , Enzo Life Sciences
Cyto-ID® Green Long-Term Cell Tracer Kit , Enzo Life Sciences
# de catalogue 10136-012
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Cyto-ID® Green Long-Term Cell Tracer Kit , Enzo Life Sciences
# de catalogue 10136-012
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    10136-012
  • Supplier No.:
    ENZ-51036-K025
Spécifications
A propos de cet article
Cyto-ID® Green long-term cell tracer kit is suitable for a variety of applications including long term cell viability, cytotoxicity, cell adhesion, cell migration and cell-cell fusion studies.
Cyto-ID® Green long-term cell tracer kit uses proprietary noncovalent cell labeling technology to stably incorporate a green fluorescent dye containing hydrophobic aliphatic chains into the cell membrane’s lipid bilayer. The dye may be loaded into cells by following the included protocol. The labeling buffer is isotonic for mammalian cells and contains no detergents or organic solvents. The appearance of labeled cells may vary depending upon the cell type from uniformly bright to punctuate. This difference is thought to relate to the extent of membrane internalization occurring after cell labeling. The Cyto-ID® Green tracer dye fluorescence is independent of pH within normally encountered physiologic ranges and fluorescence intensity per cell is typically unaffected by the ultimate pattern of dye distribution. The Cyto-ID® Green tracer dye is not toxic to cells, as determined using the benchmark MTT cell viability assay. The dye is well retained by cells for up to 96 hours after loading, and is passed to daughter cells upon mitosis.
Since the dye does not covalently modify proteins within the cells, normal physiological responses are better preserved than with molecular probes based upon thiol-reactive chloromethyl-based or amine-reactive succinimidyl ester-based fluorescent dyes. Dual labeling is also possible using a variety of available CELLestial® dyes. Labeled cells can be visualized by epifluorescence or confocal fluorescence microscopy. Additionally, dye-labeled and unlabeled cell populations can be analyzed by flow cytometry. No transfer of fluorescence to adjacent cells was observed after a prolonged 96 hour incubation period. This is in stark contrast to Calcein AM and BCECF AM, which are only retained within viable cells for a few hours at physiological temperatures. The kit is suitable for a variety of applications including long term cell viability, cytotoxicity, cell adhesion, cell migration and cell-cell fusion studies.
Ordering information: Contains Cyto-ID® Green tracer dye, 50µl4X Labeling Buffer, 12.5ml10X HBSS, 25ml.
Customers Who Bought This Also Bought