ContactUsLinkComponent
SCREEN-WELL® Purinergic Ligand Library, Enzo Life Sciences
Catalogue # 89156-286
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
SCREEN-WELL® Purinergic Ligand Library, Enzo Life Sciences
Catalogue # 89156-286
Fournisseur:  Enzo Life Sciences
Supplier Number:  BML-2820-0500
Numéro CAS:  
Spécifications
  • Size:
    1 Library
  • Cat. no.:
    89156-286
  • Supplier No.:
    2820-0500
Spécifications
À propos de cet article
The Screen-Well Purinergic and Adenosine Ligands Plate contains 72 CNS receptor ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Purinergic and Adenosine Ligands Plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.