SCREEN-WELL® Metabotropic Glutamatergic Ligand Library, Enzo Life Sciences
Catalogue # 89156-282
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Size:1 Library
- Cat. no.:89156-282
- Supplier No.:2818-0500
Spécifications
À propos de cet article
The Metabotropic Glutamatergic Ligands Plate contains 49 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Metabotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.