ContactUsLinkComponent
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
# de catalogue 89156-278
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
# de catalogue 89156-278
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Library
  • Cat. no.:
    89156-278
  • Supplier No.:
    2816-0500
Spécifications
A propos de cet article
The Histaminergic and Melatonin Ligands Plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 680 ligands.
Customers Who Bought This Also Bought