Order Entry
ContactUsLinkComponent
Bacto™ Protease Peptone No. 2, Life Technologies
Bacto™ Protease Peptone No. 2, Life Technologies
# de catalogue CA90000-492
Numéro CAS:  
Medium Proteose Peptone No 2 Bacto 10Kg
Bacto™ Protease Peptone No. 2, Life Technologies
# de catalogue CA90000-492
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Proteose Peptone, No. 2
  • Size:
    10 kg
  • Cat. no.:
    CA90000-492
  • Supplier No.:
    212110

Spécifications

A propos de cet article

Bacto™ Proteose Peptone No

  • Provides relatively high proteose content
  • Used to produce toxin-based vaccines
  • Contains only porcine origin protein

Bacto™ Proteose Peptone No. 2 is an animal-origin, enzymatic digest of protein. Similar to Bacto™ Proteose Peptone, this basic material provides relatively high proteose content and varying concentrations of nutrients, vitamins, minerals, peptides of varying sizes, buffering capabilities, and nitrogen content. However, Bacto™ Proteose Peptone No. 2 contains only porcine origin protein. Customers have successfully used this supplement to produce toxin-based vaccines, such as Bordetella pertusis for human health and other animal health vaccines.

2 is an animal-origin, enzymatic digest of protein.

Customers Who Bought This Also Bought