VACUUBRAND® XS-Series Dual-Stage Rotary Vane Vacuum Pumps for Chemistry Applications, BrandTech®
Fournisseur: Brandtech
Certificats
A propos de cet article
Two-stage rotary vane vacuum pumps, designed from the ground up for Chemistry applications
Compact design for a small footprint. Whisper quiet, and extremely low vibration. Energy efficient dual-voltage (100-120V/200-230V, 50/60Hz) motors.
Exceptional water vapor tolerance with minimal impact on ultimate vacuum. Internal steel surfaces are nitrogen-plasma treated for chemical resistance and mechanical hardness.
Certifications: NRTL tested.
Ordering information: Supplied complete, with operating manual and US plug.
Spécifications
- Catalog No:
- 10160-722
- Ultimate vacuum:
- 1.5 x 10-3 Torr (2 x 10-3 millibar)