ContactUsLinkComponent
PGE2 FPIA Kit, Enzo Life Sciences
PGE2 FPIA Kit, Enzo Life Sciences
# de catalogue 89141-756
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
PGE2 FPIA Kit 100 TESTS
PGE2 FPIA Kit, Enzo Life Sciences
# de catalogue 89141-756
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sensitivity:
    684 pg/mL (range 1,562 - 100,000 pg/mL)
  • Cat. no.:
    89141-756
  • Supplier No.:
    920-001
  • No. of Tests:
    100 Tests
Spécifications
A propos de cet article
For the quantitative determination of PGE2 in culture supernatants from any species.
The PGE2 FPIA kit is a homogeneous mix-and-read fluorescence polarization enzyme immunoassay kit with. Results are obtained within just 30 minutes and the generated signal is stable for up to 20 hours. The PGE2 FPIA kit is ideal for high throughput screening (HTS) applications. A convenient homogeneous mix-and-read assay that is amendable to miniaturization and automation, this kit has a robust Z factor >0.8.
Ordering information: Contains Conjugate concentrate, Antibody, Assay buffer concentrate, Standard.
Customers Who Bought This Also Bought