ContactUsLinkComponent
Nitric Oxide (NO2-/NO3-) Detection Kit, Enzo Life Sciences
Nitric Oxide (NO2-/NO3-) Detection Kit, Enzo Life Sciences
# de catalogue 89141-752
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
NITRIC OXIDE (NO2/NO3) Detection Kit 2 X 96 Wells
Nitric Oxide (NO2-/NO3-) Detection Kit, Enzo Life Sciences
# de catalogue 89141-752
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sensitivity:
    (Nitrite) 0.222 µmole/L; (Nitrate) 0.625 µmole/L (range (Nitrite) 3.125 - 200 µmole/L; (Nitrate) 3.125 - 100 µmole/L)
  • Cat. no.:
    89141-752
  • Supplier No.:
    917-010
  • No. of Tests:
    96 Wells
Spécifications
A propos de cet article
For the quantitative determination of nitrite and nitrate levels in culture supernatants, plasma, serum, saliva, and urine from any species.
The Nitric Oxide (NO2-/NO3-), detection kit is a complete kit for the quantitative determination of nitrate and nitrite concentrations. Absorbance is read at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.
Ordering information: Contains Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Nitrite Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II.
Customers Who Bought This Also Bought