Spécifications
- Sensitivity:16.4 Units/mL (range 76 - 933 Units/mL)
- No. of wells:96 Wells
- Cat. no.:89141-670
- Supplier No.:907-013
Spécifications
A propos de cet article
For the colorimetric measurement of Caspase-3 in cell lysates and buffers from any species.
The Caspase-3 colorimetric detection kit provides a higher throughput method for measuring caspase-3 activity in up 37 samples in duplicate per kit. The kit is flexible, providing both kinetic or end-point assay options.