ContactUsLinkComponent
Caspase-3 Colorimetric Detection Kit, Enzo Life Sciences
Caspase-3 Colorimetric Detection Kit, Enzo Life Sciences
# de catalogue 89141-670
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
CASPASE-3 COLORIMETRIC Detection Kit 1 X 96 Wells
Caspase-3 Colorimetric Detection Kit, Enzo Life Sciences
# de catalogue 89141-670
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sensitivity:
    16.4 Units/mL (range 76 - 933 Units/mL)
  • No. of wells:
    96 Wells
  • Cat. no.:
    89141-670
  • Supplier No.:
    907-013
Spécifications
A propos de cet article
For the colorimetric measurement of Caspase-3 in cell lysates and buffers from any species.
The Caspase-3 colorimetric detection kit provides a higher throughput method for measuring caspase-3 activity in up 37 samples in duplicate per kit. The kit is flexible, providing both kinetic or end-point assay options.
Ordering information: Contains Half-Area Microtiter plate, Standard, DTT, p-Nitroaniline Calibrator, Reaction buffer concentrate, Substrate concentrate, Stop solution 2.
Customers Who Bought This Also Bought