Hydrogen Peroxide Chemiluminescent Detection Kit, Enzo Life Sciences
# de catalogue 89141-668
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Sensitivity:2.39 ng/mL (range 2.44 - 2,500 ng/mL)
- No. of wells:96 Wells
- Cat. no.:89141-668
- Supplier No.:907-012
Spécifications
A propos de cet article
For the quantitative determination of hydrogen peroxide in culture supernatants and cell lysates from any species.
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit but is not recommended for samples that contain phenol red or hemoglobin.
Ordering information: Contains White Microtiter plate, Standard, Substrate, Trigger solution.
Customers Who Bought This Also Bought